siRNA Details
| Gene Symbol: POSTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135934.1 | NP_001129406.1 | 6 | 764 | ||
| NM_001135935.1 | NP_001129407.1 | 6 | 764 | |||
| NM_001135936.1 | NP_001129408.1 | 6 | 764 | |||
| NM_001286665.1 | NP_001273594.1 | 6 | 764 | |||
| NM_001286666.1 | NP_001273595.1 | 6 | 764 | |||
| NM_001286667.1 | NP_001273596.1 | 6 | 764 | |||
| NM_006475.2 | NP_006466.2 | 6 | 764 | |||
| XM_005266231.2 | XP_005266288.1 | 6 | 759 | |||
| XM_005266232.2 | XP_005266289.1 | 6 | 759 | |||
| XM_017020355.1 | XP_016875844.1 | 6 | 759 | |||
| XM_017020356.1 | XP_016875845.1 | 6 | 759 | |||
| GenBank mRNA | AK300261.1 | - | 6 | 878 | ||
| AY140646.1 | - | 6 | 657 | |||
| AY918092.1 | - | 6 | 657 | |||
| BC106709.1 | - | 6 | 657 | 40007999 | ||
| BC106710.1 | - | 6 | 657 | 40008000 | ||
| BY795054.2 | - | 6 | 671 | |||
| D13665.1 | - | 6 | 673 | |||
| D13666.1 | - | 6 | 657 | |||
| EU262883.1 | - | 6 | 646 | |||
| EU262884.1 | - | 6 | 646 | |||
| EU262885.1 | - | 6 | 646 | |||
| EU262886.1 | - | 6 | 646 | |||
| GU354210.1 | - | 6 | 657 | |||

POSTN
Genomic Map