siRNA Details
| Gene Symbol: MTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006635.2 | NP_001006636.1 | 6 | 680 | ||
| NM_001319097.1 | NP_001306026.1 | 5 | 476 | |||
| NM_001319098.1 | NP_001306027.1 | 5 | 476 | |||
| NM_006554.4 | NP_006545.1 | 5 | 476 | |||
| XM_011510497.2 | XP_011508799.1 | 6 | 573 | |||
| GenBank mRNA | AF053551.1 | - | 5 | 449 | ||
| AK289359.1 | - | 5 | 427 | |||
| AK303564.1 | - | 6 | 605 | |||
| BC017271.2 | - | 6 | 610 | 4635884 | ||
| BC067831.1 | - | 5 | 455 | 5298149 | ||
| BC088359.1 | - | 5 | 411 | 30378040 | ||
| BG699784.1 | - | 6 | 684 | |||
| BU165147.1 | - | 5 | 407 | |||
| CX165048.1 | - | 5 | 369 | |||

MTX2
Genomic Map