siRNA Details
| Gene Symbol: C6orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286474.1 | NP_001273403.1 | 26 | 1201 | ||
| NM_001286475.1 | NP_001273404.1 | 24 | 1159 | |||
| NM_006781.4 | NP_006772.3 | 23 | 1207 | |||
| XM_017010182.1 | XP_016865671.1 | 16 | 728 | |||
| XM_017010185.1 | XP_016865674.1 | 24 | 1232 | |||
| GenBank mRNA | AK225742.1 | - | 26 | 1192 | ||
| AK302176.1 | - | 24 | 1119 | |||
| AK310230.1 | - | 25 | 1169 | |||
| BC034774.1 | - | 23 | 1174 | 5169157 | ||

C6orf10
Genomic Map