siRNA Details
| Gene Symbol: CD226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303618.1 | NP_001290547.1 | 3 | 966 | ||
| NM_001303619.1 | NP_001290548.1 | 2 | 559 | |||
| NM_006566.3 | NP_006557.2 | 4 | 968 | |||
| XM_005266642.3 | XP_005266699.1 | 4 | 1067 | |||
| XM_005266643.3 | XP_005266700.1 | 2 | 161 | |||
| XM_006722374.3 | XP_006722437.1 | 2 | 632 | |||
| XM_017025524.1 | XP_016881013.1 | 4 | 7711 | |||
| XM_017025525.1 | XP_016881014.1 | 2 | 443 | |||
| XM_017025526.1 | XP_016881015.1 | 3 | 337 | |||
| XM_017025527.1 | XP_016881016.1 | 2 | 1001 | |||
| GenBank mRNA | AJ537553.1 | - | 3 | 548 | ||
| AK313199.1 | - | 3 | 967 | |||
| BC074787.2 | - | 3 | 553 | 30915562 | ||
| GU935335.1 | - | 4 | 715 | |||
| GU935336.1 | - | 2 | 318 | |||
| U56102.1 | - | 4 | 727 | |||

CD226
Genomic Map