siRNA Details
| Gene Symbol: STAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042749.2 | NP_001036214.1 | 30 | 3597 | ||
| NM_001042750.1 | NP_001036215.1 | 30 | 3441 | |||
| NM_001042751.1 | NP_001036216.1 | 30 | 3441 | |||
| NM_001282418.1 | NP_001269347.1 | 30 | 3420 | |||
| NM_006603.4 | NP_006594.3 | 29 | 3376 | |||
| XM_005262357.2 | XP_005262414.1 | 29 | 3466 | |||
| XM_005262358.2 | XP_005262415.1 | 30 | 3403 | |||
| XM_005262359.3 | XP_005262416.1 | 29 | 3356 | |||
| XM_005262360.2 | XP_005262417.1 | 29 | 3285 | |||
| XM_005262361.2 | XP_005262418.1 | 29 | 3413 | |||
| XM_006724727.1 | XP_006724790.1 | 31 | 3545 | |||
| XM_006724728.3 | XP_006724791.1 | 30 | 3479 | |||
| XM_011531253.2 | XP_011529555.1 | 29 | 3338 | |||
| XM_017029232.1 | XP_016884721.1 | 30 | 3403 | |||
| XM_017029233.1 | XP_016884722.1 | 29 | 3285 | |||
| XM_017029234.1 | XP_016884723.1 | 29 | 3338 | |||
| XM_017029235.1 | XP_016884724.1 | 30 | 3484 | |||
| GenBank mRNA | AK298633.1 | - | 19 | 2095 | ||
| AK313559.1 | - | 29 | 3268 | |||
| AL831939.2 | - | 29 | 3344 | |||
| BC001765.1 | - | 29 | 3325 | 3352347 | ||
| BX640970.1 | - | 30 | 3452 | |||
| BX641002.1 | - | 30 | 3345 | |||
| BX641003.1 | - | 30 | 3410 | |||
| CR627425.1 | - | 30 | 3419 | |||
| CR749433.1 | - | 30 | 3410 | |||

STAG2
Genomic Map