siRNA Details
| Gene Symbol: SIX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016932.4 | NP_058628.3 | 1 | 557 | ||
| XM_005264100.3 | XP_005264157.1 | 1 | 799 | |||
| GenBank mRNA | AF136940.1 | - | 1 | 316 | ||
| AF332196.1 | - | 1 | 254 | |||
| AF332197.1 | - | 1 | 557 | |||
| BC024033.2 | - | 1 | 537 | 3027992 | ||
| BT020100.1 | - | 1 | 264 | |||

SIX2
Genomic Map