siRNA Details
| Gene Symbol: HBS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145158.1 | NP_001138630.1 | 11 | 1530 | ||
| NM_006620.3 | NP_006611.1 | 12 | 1656 | |||
| GenBank mRNA | AB028961.1 | - | 7 | 885 | ||
| AJ459826.1 | - | 12 | 1642 | |||
| AK024258.1 | - | 12 | 1577 | |||
| AK293573.1 | - | 9 | 864 | |||
| AK295545.1 | - | 11 | 1451 | |||
| AK298336.1 | - | 9 | 1059 | |||
| BC001465.2 | - | 12 | 1565 | 3138910 | ||
| BC040849.1 | - | 12 | 1560 | 5104748 | ||
| BX648033.1 | - | 9 | 1321 | |||
| U87791.1 | - | 12 | 1643 | |||

HBS1L
Genomic Map