siRNA Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006635.3 | NP_006626.3 | 3 | 1943 | ||
| XM_005258422.3 | XP_005258479.1 | 3 | 2308 | |||
| XM_017026176.1 | XP_016881665.1 | 3 | 1758 | |||
| GenBank mRNA | AK090816.1 | - | 1 | 701 | ||
| AK292144.1 | - | 3 | 1943 | |||
| AK298106.1 | - | 3 | 1665 | |||
| AY329492.1 | - | 3 | 1864 | |||
| BC109018.1 | - | 3 | 1621 | |||
| BC109019.1 | - | 3 | 1621 | |||
| BC118625.1 | - | 3 | 1621 | 40006201 | ||
| X78931.1 | - | 1 | 457 | |||

ZNF460
Genomic Map