siRNA Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286132.1 | NP_001273061.1 | 5, 6 | 739 | ||
| NM_001286133.1 | NP_001273062.1 | 5, 6 | 682 | |||
| NM_006638.3 | NP_006629.2 | 6, 7 | 808 | |||
| XM_011514253.2 | XP_011512555.1 | 6, 7 | 803 | |||
| GenBank mRNA | BC017871.1 | - | 5, 6 | 713 | 4278825 | |
| BU607286.1 | - | 2, 3 | 389 | |||

RPP40
Genomic Map