siRNA Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286132.1 | NP_001273061.1 | 7 | 954 | ||
| NM_001286133.1 | NP_001273062.1 | 7 | 897 | |||
| NM_006638.3 | NP_006629.2 | 8 | 1023 | |||
| GenBank mRNA | BC017871.1 | - | 7 | 928 | 4278825 | |
| BU607286.1 | - | 1 | 174 | |||

RPP40
Genomic Map