siRNA Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166221.1 | NP_001159693.1 | 10 | 905 | ||
| NM_006649.3 | NP_006640.2 | 11 | 1061 | |||
| XM_017029237.1 | XP_016884726.1 | 13 | 1213 | |||
| XM_017029238.1 | XP_016884727.1 | 11 | 1243 | |||
| GenBank mRNA | AK094408.1 | - | 2 | 57 | ||
| AK291918.1 | - | 11 | 1028 | |||
| AK298578.1 | - | 10 | 874 | |||
| AK304563.1 | - | 10 | 926 | |||
| BC001149.1 | - | 11 | 997 | 3355662 | ||
| BC009649.1 | - | 11 | 999 | 3459192 | ||
| BC014987.1 | - | 11 | 989 | 4901477 | ||

UTP14A
Genomic Map