siRNA Details
| Gene Symbol: TSPAN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168320.1 | NP_001161792.1 | 8 | 820 | ||
| NM_006675.4 | NP_006666.1 | 9 | 887 | |||
| XM_011520912.2 | XP_011519214.1 | 10 | 1004 | |||
| XM_011520913.2 | XP_011519215.1 | 7 | 746 | |||
| GenBank mRNA | AF089749.1 | - | 9 | 830 | ||
| AK027366.1 | - | 9 | 863 | |||
| AK223179.1 | - | 9 | 859 | |||
| BC071881.1 | - | 8 | 802 | 6256950 | ||
| CR542109.1 | - | 7 | 704 | |||

TSPAN9
Genomic Map