siRNA Details
| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 25 | 2608 | ||
| GenBank mRNA | AB209201.1 | - | 13 | 1113 | ||
| AF083217.1 | - | 25 | 2589 | |||
| AK022858.1 | - | 17 | 1613 | |||
| AK292438.1 | - | 25 | 2603 | |||

WDR3
Genomic Map