siRNA Details
| Gene Symbol: LYVE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006691.3 | NP_006682.2 | 4 | 700 | ||
| GenBank mRNA | AF127670.2 | - | 4 | 534 | ||
| AK075443.1 | - | 4 | 614 | |||
| AK312462.1 | - | 4 | 698 | |||
| AY358925.1 | - | 4 | 573 | |||
| BC026231.1 | - | 4 | 459 | 4712863 | ||

LYVE1
Genomic Map