siRNA Details
| Gene Symbol: SRSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032102.3 | NP_115285.1 | 1 | 919 | ||
| NR_103726.1 | 1 | 919 | ||||
| XM_017017142.1 | XP_016872631.1 | 1 | 881 | |||
| GenBank mRNA | AF031166.1 | - | 1 | 539 | ||
| AK023379.1 | - | 1 | 805 | |||
| AK312514.1 | - | 1 | 808 | |||
| BC057783.1 | - | 2 | 609 | 5285882 | ||
| CV805885.1 | - | 1 | 302 | |||

SRSF8
Genomic Map