siRNA Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130914.1 | NP_001124386.1 | 2 | 345 | ||
| NM_006806.4 | NP_006797.3 | 2 | 345 | |||
| XM_011529441.1 | XP_011527743.1 | 3 | 453 | |||
| XM_017028265.1 | XP_016883754.1 | 3 | 483 | |||
| XM_017028266.1 | XP_016883755.1 | 3 | 440 | |||
| GenBank mRNA | AK289408.1 | - | 2 | 293 | ||
| BC011957.1 | - | 2 | 239 | 3859781 | ||
| BT007276.1 | - | 1 | 85 | |||
| CR457061.1 | - | 1 | 85 | |||
| D64110.1 | - | 2 | 179 | |||
| DA381239.1 | - | 2 | 346 | |||
| DR155504.1 | - | 2 | 261 | |||

BTG3
Genomic Map