siRNA Details
| Gene Symbol: PNRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006813.2 | NP_006804.1 | 2 | 980 | ||
| GenBank mRNA | AK130046.1 | - | 2 | 938 | ||
| AK130722.1 | - | 1 | 1020 | |||
| AK312665.1 | - | 2 | 939 | |||
| AY303779.1 | - | 2 | 558 | |||
| BC018112.1 | - | 2 | 850 | 3916954 | ||
| BC044919.1 | - | 2 | 1160 | |||
| U03105.1 | - | 2 | 908 | |||

PNRC1
Genomic Map