siRNA Details
| Gene Symbol: LMAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006816.2 | NP_006807.1 | 2, 3 | 563 | ||
| GenBank mRNA | AK222609.1 | - | 2, 3 | 330 | ||
| AK292099.1 | - | 2, 3 | 340 | |||
| AK308027.1 | - | 2, 3 | 474 | |||
| BC017263.2 | - | 2, 3 | 343 | 4135210 | ||
| DC404202.1 | - | 2, 3 | 563 | |||
| U10362.1 | - | 2, 3 | 326 | |||

LMAN2
Genomic Map