siRNA Details
| Gene Symbol: SLC27A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317929.1 | NP_001304858.1 | 2 | 1015 | ||
| NM_024330.1 | NP_077306.1 | 2 | 1015 | |||
| GenBank mRNA | AJ577572.1 | - | 2 | 1308 | ||
| AK027499.1 | - | 2 | 919 | |||
| AK074134.1 | - | 1 | 1341 | |||
| AK075377.1 | - | 2 | 1008 | |||
| AK123997.1 | - | 3 | 479 | |||
| AK125102.1 | - | 2 | 1953 | |||
| AY358409.1 | - | 2 | 1003 | |||
| BC003041.1 | - | 2 | 1015 | 2822875 | ||
| BC003654.2 | - | 1 | 64 | |||
| BC009916.2 | - | 2 | 1015 | 2822875 | ||
| BC029792.1 | - | 2 | 883 | 5195359 | ||

SLC27A3
Genomic Map