siRNA Details
| Gene Symbol: GLIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006851.2 | NP_006842.2 | 2 | 496 | ||
| XM_011537775.2 | XP_011536077.1 | 2 | 475 | |||
| XM_017018721.1 | XP_016874210.1 | 2 | 376 | |||
| GenBank mRNA | AF400440.1 | - | 2 | 376 | ||
| AK304847.1 | - | 2 | 476 | |||
| AK313449.1 | - | 2 | 362 | |||
| BC012510.1 | - | 2 | 412 | 4453841 | ||
| CB123261.1 | - | 2 | 549 | |||
| EF667354.1 | - | 2 | 348 | |||
| U16307.3 | - | 2 | 432 | |||
| X91911.1 | - | 2 | 476 | |||

GLIPR1
Genomic Map