siRNA Details
| Gene Symbol: LILRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130917.2 | NP_001124389.2 | 4 | 493 | ||
| NM_001290270.1 | NP_001277199.1 | 3 | 457 | |||
| NM_001290271.1 | NP_001277200.1 | 5 | 932 | |||
| NM_006866.3 | NP_006857.2 | 5 | 1065 | |||
| XM_006722986.1 | XP_006723049.1 | 5 | 1065 | |||
| XM_011526385.1 | XP_011524687.1 | 5 | 1065 | |||
| XM_011526386.1 | XP_011524688.1 | 4 | 1029 | |||
| XM_011526387.1 | XP_011524689.1 | 4 | 1029 | |||
| XM_011526388.1 | XP_011524690.1 | 4 | 1029 | |||
| XM_011526389.1 | XP_011524691.1 | 3 | 747 | |||
| XM_011526390.1 | XP_011524692.1 | 5 | 1065 | |||
| XM_011526391.1 | XP_011524693.1 | 5 | 1065 | |||
| XM_017026224.1 | XP_016881713.1 | 5 | 1065 | |||
| GenBank mRNA | AF025531.1 | - | 4 | 484 | ||
| AK130170.1 | - | 4 | 523 | |||
| AK222651.1 | - | 4 | 489 | |||
| AK310088.1 | - | 5 | 914 | |||
| BC017412.1 | - | 4 | 495 | 4618777 | ||
| BC027916.1 | - | 4 | 478 | 5223154 | ||
| BP296918.1 | - | 4 | 534 | |||
| EU915609.1 | - | 3 | 426 | |||
| EU915610.1 | - | 4 | 462 | |||
| U82275.1 | - | 4 | 532 | |||
| U82276.1 | - | 3 | 447 | |||
| U82278.1 | - | 4 | 445 | |||

LILRA2
Genomic Map