siRNA Details
| Gene Symbol: RAB31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006868.3 | NP_006859.2 | 5 | 537 | ||
| XM_017025528.1 | XP_016881017.1 | 6 | 919 | |||
| XM_017025529.1 | XP_016881018.1 | 6 | 907 | |||
| GenBank mRNA | AF183421.1 | - | 5 | 490 | ||
| AF234995.1 | - | 5 | 536 | |||
| AF498957.1 | - | 5 | 359 | |||
| AK223505.1 | - | 5 | 401 | |||
| AK314809.1 | - | 5 | 495 | |||
| BC001148.1 | - | 5 | 459 | 3534853 | ||
| BT020027.1 | - | 5 | 359 | |||
| BT020028.1 | - | 5 | 359 | |||
| BU944638.1 | - | 5 | 603 | |||
| CR407659.1 | - | 5 | 359 | |||
| U57091.1 | - | 5 | 404 | |||
| U59877.1 | - | 5 | 419 | |||

RAB31
Genomic Map