siRNA Details
| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006875.3 | NP_006866.2 | 6 | 1167 | ||
| GenBank mRNA | AK290931.1 | - | 6 | 1055 | ||
| AK312745.1 | - | 6 | 1055 | |||
| BC018111.1 | - | 6 | 1038 | 3913552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIM2| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006875.3 | NP_006866.2 | 6 | 1167 | ||
| GenBank mRNA | AK290931.1 | - | 6 | 1055 | ||
| AK312745.1 | - | 6 | 1055 | |||
| BC018111.1 | - | 6 | 1038 | 3913552 | ||