siRNA Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012216.3 | NP_036348.2 | 2 | 1267 | ||
| NM_052817.2 | NP_438112.2 | 2 | 1267 | |||
| XM_005262062.4 | XP_005262119.1 | 2 | 5530 | |||
| XM_017029239.1 | XP_016884728.1 | 2 | 5530 | |||
| GenBank mRNA | AF196481.1 | - | 2 | 707 | ||
| AY625004.1 | - | 1 | 634 | |||
| BC017707.1 | - | 1 | 661 | 4446324 | ||
| BT006663.1 | - | 1 | 634 | |||
| Y18880.1 | - | 2 | 891 | |||

MID2
Genomic Map