siRNA Details
| Gene Symbol: CYB561D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291284.1 | NP_001278213.1 | 4 | 920 | ||
| NM_007022.4 | NP_008953.1 | 4 | 986 | |||
| GenBank mRNA | AF040704.1 | - | 4 | 743 | ||
| AK291167.1 | - | 4 | 806 | |||
| BC047691.1 | - | 4 | 815 | 6060648 | ||

CYB561D2
Genomic Map