siRNA Details
| Gene Symbol: RER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007033.4 | NP_008964.3 | 3 | 345 | ||
| XM_005244713.4 | XP_005244770.2 | 3 | 1166 | |||
| XM_006710306.2 | XP_006710369.1 | 4 | 427 | |||
| XM_011540542.1 | XP_011538844.1 | 4 | 365 | |||
| XM_011540543.1 | XP_011538845.1 | 3 | 213 | |||
| XM_017000131.1 | XP_016855620.1 | 3 | 1696 | |||
| XM_017000132.1 | XP_016855621.1 | 4 | 455 | |||
| XM_017000133.1 | XP_016855622.1 | 4 | 538 | |||
| XM_017000134.1 | XP_016855623.1 | 5 | 452 | |||
| XM_017000135.1 | XP_016855624.1 | 3 | 452 | |||
| GenBank mRNA | AF091071.1 | - | 3 | 230 | ||
| AF157324.1 | - | 3 | 284 | |||
| AJ001421.1 | - | 2 | 154 | |||
| AK095750.1 | - | 2 | 1940 | |||
| AK289658.1 | - | 3 | 279 | |||
| BC004965.1 | - | 3 | 280 | 3546970 | ||
| BT007224.1 | - | 2 | 154 | |||
| CR456806.1 | - | 2 | 154 | |||
| DA409924.1 | - | 4 | 427 | |||

RER1
Genomic Map