siRNA Details
| Gene Symbol: ADAM30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021794.3 | NP_068566.2 | 1 | 870 | ||
| GenBank mRNA | AF171932.1 | - | 1 | 682 | ||
| AF171933.1 | - | 1 | 682 | |||
| AK292483.1 | - | 1 | 861 | |||
| AY358734.1 | - | 1 | 844 | |||
| BC028372.2 | - | 1 | 841 | 4831258 | ||

ADAM30
Genomic Map