siRNA Details
| Gene Symbol: KATNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204076.1 | NP_001191005.1 | 7 | 1187 | ||
| NM_007044.3 | NP_008975.1 | 9 | 1415 | |||
| XM_005266804.2 | XP_005266861.1 | 9 | 1195 | |||
| XM_005266805.3 | XP_005266862.1 | 8 | 1175 | |||
| XM_017010207.1 | XP_016865696.1 | 10 | 1505 | |||
| XM_017010208.1 | XP_016865697.1 | 8 | 1231 | |||
| XM_017010209.1 | XP_016865698.1 | 8 | 1073 | |||
| XM_017010210.1 | XP_016865699.1 | 9 | 1200 | |||
| GenBank mRNA | AF056022.1 | - | 8 | 1111 | ||
| AK292090.1 | - | 9 | 1325 | |||
| BC050428.1 | - | 7 | 1024 | 5764501 | ||
| CA426364.1 | - | 3 | 491 | |||

KATNA1
Genomic Map