siRNA Details
| Gene Symbol: GLMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319683.1 | NP_001306612.1 | 2 | 128 | ||
| NM_053274.2 | NP_444504.1 | 2 | 128 | |||
| NR_135089.1 | 2 | 128 | ||||
| XM_005270401.3 | XP_005270458.1 | 2 | 116 | |||
| XM_006710309.2 | XP_006710372.1 | 2 | 106 | |||
| XM_011540546.2 | XP_011538848.1 | 4 | 291 | |||
| XM_017000137.1 | XP_016855626.1 | 2 | 496 | |||
| XM_017000138.1 | XP_016855627.1 | 2 | 496 | |||
| XM_017000139.1 | XP_016855628.1 | 2 | 496 | |||
| XM_017000140.1 | XP_016855629.1 | 2 | 232 | |||
| XM_017000141.1 | XP_016855630.1 | 2 | 128 | |||
| XM_017000142.1 | XP_016855631.1 | 2 | 106 | |||
| XM_017000143.1 | XP_016855632.1 | 2 | 487 | |||
| XM_017000144.1 | XP_016855633.1 | 2 | 93 | |||
| GenBank mRNA | AJ302735.1 | - | 2 | 94 | ||
| AJ347709.1 | - | 2 | 95 | |||
| AK295971.1 | - | 2 | 106 | |||
| BC001257.1 | - | 2 | 95 | 3454182 | ||
| DA729622.1 | - | 2 | 128 | |||
| U73704.1 | - | 2 | 106 | |||

GLMN
Genomic Map