siRNA Details
| Gene Symbol: HHLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031693.2 | NP_001026863.1 | 1 | 159 | ||
| NM_001036645.1 | NP_001031722.1 | 1 | 159 | |||
| NM_001036646.1 | NP_001031723.1 | 1 | 159 | |||
| NR_027404.1 | 1 | 159 | ||||
| XM_011540547.2 | XP_011538849.1 | 1 | 1129 | |||
| XM_011540548.2 | XP_011538850.1 | 1 | 54 | |||
| GenBank mRNA | AF126164.1 | - | 1 | 140 | ||
| BC010922.1 | - | 1 | 137 | 4064190 | ||
| BG773961.1 | - | 1 | 159 | |||
| BI868263.1 | - | 1 | 159 | |||

HHLA3
Genomic Map