siRNA Details
| Gene Symbol: AP4S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128126.2 | NP_001121598.1 | 4, 5 | 677 | ||
| NM_001254727.1 | NP_001241656.1 | 4, 5 | 677 | |||
| NM_001254728.1 | NP_001241657.1 | 4, 5 | 1034 | |||
| NM_001254729.1 | NP_001241658.1 | 4, 5 | 552 | |||
| NM_007077.4 | NP_009008.2 | 4, 5 | 677 | |||
| XM_005267293.4 | XP_005267350.1 | 5, 6 | 1120 | |||
| XM_011536371.2 | XP_011534673.1 | 5, 6 | 690 | |||
| XM_011536372.2 | XP_011534674.1 | 4, 5 | 1084 | |||
| GenBank mRNA | AB030654.1 | - | 4, 5 | 497 | ||
| AF155159.1 | - | 4, 5 | 471 | |||
| AK304115.1 | - | 4, 5 | 498 | |||
| AK316517.1 | - | 4, 5 | 500 | |||
| BC001259.1 | - | 4, 5 | 460 | 3454117 | ||
| BT006701.1 | - | 3, 4 | 288 | |||
| BX247969.1 | - | 4, 5 | 479 | |||
| CR457100.1 | - | 3, 4 | 288 | |||

AP4S1
Genomic Map