siRNA Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007080.2 | NP_009011.1 | 3, 4 | 358 | ||
| XM_017007672.1 | XP_016863161.1 | 3, 4 | 333 | |||
| XM_017007673.1 | XP_016863162.1 | 3, 4 | 658 | |||
| GenBank mRNA | AF182292.1 | - | 3, 4 | 300 | ||
| AI351504.1 | - | 1 | 428 | |||
| AJ238098.1 | - | 3, 4 | 300 | |||
| AK312126.1 | - | 3, 4 | 293 | |||
| BC016026.1 | - | 3, 4 | 312 | 3910698 | ||
| BC019894.1 | - | 3, 4 | 938 | |||
| DB465201.1 | - | 3, 4 | 359 | |||

LSM6
Genomic Map