siRNA Details
| Gene Symbol: ADAMTS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197941.3 | NP_922932.2 | 15 | 2763 | ||
| NR_135689.1 | 16 | 2914 | ||||
| XM_011543113.2 | XP_011541415.1 | 15 | 2778 | |||
| XM_011543114.2 | XP_011541416.1 | 15 | 2357 | |||
| XM_011543115.2 | XP_011541417.1 | 14 | 2666 | |||
| XM_011543116.2 | XP_011541418.1 | 13 | 2525 | |||
| XM_011543117.1 | XP_011541419.1 | 13 | 2116 | |||
| XM_011543119.2 | XP_011541421.1 | 15 | 2807 | |||
| XM_011543120.2 | XP_011541422.1 | 15 | 2804 | |||
| XM_011543121.2 | XP_011541423.2 | 11 | 1144 | |||
| XM_011543122.2 | XP_011541424.1 | 15 | 2804 | |||
| XM_011543123.2 | XP_011541425.1 | 9 | 882 | |||
| XM_011543124.2 | XP_011541426.1 | 6 | 666 | |||
| XM_011543125.1 | XP_011541427.1 | 5 | 521 | |||
| GenBank mRNA | AB209685.1 | - | 16 | 2411 | ||
| AF140674.1 | - | 13 | 1783 | |||
| AY692424.1 | - | 15 | 2721 | |||
| AY692425.1 | - | 15 | 2062 | |||
| AY692426.1 | - | 14 | 1929 | |||

ADAMTS6
Genomic Map