siRNA Details
| Gene Symbol: ADAMTS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197941.3 | NP_922932.2 | 3 | 1139 | ||
| NR_135689.1 | 3 | 1139 | ||||
| XM_011543113.2 | XP_011541415.1 | 3 | 1154 | |||
| XM_011543114.2 | XP_011541416.1 | 3 | 733 | |||
| XM_011543115.2 | XP_011541417.1 | 3 | 1192 | |||
| XM_011543116.2 | XP_011541418.1 | 3 | 1189 | |||
| XM_011543119.2 | XP_011541421.1 | 3 | 1183 | |||
| XM_011543120.2 | XP_011541422.1 | 3 | 1180 | |||
| XM_011543122.2 | XP_011541424.1 | 3 | 1180 | |||
| XM_011543126.2 | XP_011541428.1 | 3 | 1177 | |||
| XM_011543127.2 | XP_011541429.1 | 3 | 1177 | |||
| GenBank mRNA | AB209685.1 | - | 3 | 636 | ||
| AF140674.1 | - | 2 | 303 | |||
| AY692424.1 | - | 3 | 1097 | |||
| AY692425.1 | - | 2 | 301 | |||
| AY692426.1 | - | 2 | 300 | |||

ADAMTS6
Genomic Map