siRNA Details
| Gene Symbol: BAZ1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013448.2 | NP_038476.2 | 18 | 3128 | ||
| NM_182648.1 | NP_872589.1 | 17 | 3032 | |||
| XM_011536374.1 | XP_011534676.1 | 17 | 2427 | |||
| XM_011536376.2 | XP_011534678.2 | 8 | 1255 | |||
| XM_017020942.1 | XP_016876431.1 | 7 | 1158 | |||
| GenBank mRNA | AB032252.1 | - | 17 | 3029 | ||
| AB209255.1 | - | 9 | 1379 | |||
| AF213467.1 | - | 18 | 2726 | |||
| AF221130.1 | - | 17 | 2560 | |||
| AL050089.1 | - | 1 | 325 | |||

BAZ1A
Genomic Map