siRNA Details
| Gene Symbol: ZNF277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021994.2 | NP_068834.2 | 12 | 1419 | ||
| XM_005250129.3 | XP_005250186.1 | 11 | 1221 | |||
| XM_011515768.2 | XP_011514070.1 | 12 | 1391 | |||
| XM_017011720.1 | XP_016867209.1 | 11 | 1213 | |||
| GenBank mRNA | AF209198.1 | - | 12 | 1419 | ||
| AF308819.1 | - | 12 | 1290 | |||
| AK314630.1 | - | 12 | 1307 | |||
| AL713791.1 | - | 12 | 1306 | |||

ZNF277
Genomic Map