siRNA Details
| Gene Symbol: SLC2A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017585.3 | NP_060055.2 | 8, 9 | 1307 | ||
| XM_011518189.2 | XP_011516491.2 | 8, 9 | 1284 | |||
| XM_017014237.1 | XP_016869726.1 | 8, 9 | 1275 | |||
| XM_017014238.1 | XP_016869727.1 | 8, 9 | 1279 | |||
| GenBank mRNA | AJ011372.2 | - | 8, 9 | 1249 | ||
| AK074927.1 | - | 8, 9 | 1290 | |||
| AK096630.1 | - | 6, 7 | 2184 | |||
| AK222919.1 | - | 8, 9 | 1252 | |||
| AK225106.1 | - | 8, 9 | 1180 | |||
| BC013740.1 | - | 8, 9 | 1241 | 4449594 | ||
| Y17803.1 | - | 8, 9 | 1258 | |||

SLC2A6
Genomic Map