siRNA Details
| Gene Symbol: PROSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007198.3 | NP_009129.1 | 5 | 500 | ||
| GenBank mRNA | AK021923.1 | - | 5 | 368 | ||
| AK291135.1 | - | 5 | 438 | |||
| AL136616.1 | - | 5 | 480 | |||
| BC012334.1 | - | 5 | 490 | 3546307 | ||
| CR533532.1 | - | 5 | 423 | |||
| DA086552.1 | - | 5 | 500 | |||

PROSC
Genomic Map