siRNA Details
| Gene Symbol: KLF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007249.4 | NP_009180.3 | 4 | 437 | ||
| XM_005266251.3 | XP_005266308.1 | 4 | 546 | |||
| XM_011534907.2 | XP_011533209.1 | 3 | 260 | |||
| XM_011534908.2 | XP_011533210.1 | 3 | 260 | |||
| XM_011534909.2 | XP_011533211.1 | 4 | 393 | |||
| XM_011534910.2 | XP_011533212.1 | 3 | 224 | |||
| XM_011534911.2 | XP_011533213.1 | 3 | 302 | |||
| XM_011534912.2 | XP_011533214.1 | 3 | 373 | |||
| XM_017020384.1 | XP_016875873.1 | 3 | 204 | |||
| XM_017020385.1 | XP_016875874.1 | 4 | 477 | |||
| GenBank mRNA | AF113122.1 | - | 4 | 411 | ||
| AF161471.1 | - | 4 | 285 | |||
| AF330041.1 | - | 3 | 223 | |||
| AJ243274.1 | - | 3 | 215 | |||
| AK291397.1 | - | 4 | 533 | |||
| BC019680.1 | - | 3 | 242 | 4942405 | ||
| DB125574.1 | - | 4 | 282 | |||
| HF546212.1 | - | 4 | 408 | |||

KLF12
Genomic Map