siRNA Details
| Gene Symbol: CYP4F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007253.3 | NP_009184.1 | 7 | 778 | ||
| XM_017026232.1 | XP_016881721.1 | 6 | 715 | |||
| GenBank mRNA | AF133298.1 | - | 6 | 721 | ||
| AK096809.1 | - | 3 | 2432 | |||
| AK300530.1 | - | 6 | 869 | |||
| AK300539.1 | - | 3 | 329 | |||
| AK314685.1 | - | 7 | 776 | |||

CYP4F8
Genomic Map