siRNA Details
| Gene Symbol: RBPJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281448.1 | NP_001268377.1 | 8, 9 | 953 | ||
| NM_001281449.1 | NP_001268378.1 | 8, 9 | 953 | |||
| NM_014276.3 | NP_055091.2 | 8, 9 | 953 | |||
| XM_011528521.2 | XP_011526823.1 | 8, 9 | 960 | |||
| XM_011528522.2 | XP_011526824.1 | 8, 9 | 960 | |||
| XM_011528523.2 | XP_011526825.1 | 8, 9 | 960 | |||
| GenBank mRNA | AB024964.1 | - | 8, 9 | 873 | ||
| AB026048.1 | - | 8, 9 | 873 | |||
| AB027710.1 | - | 8, 9 | 873 | |||

RBPJL
Genomic Map