siRNA Details
| Gene Symbol: ECD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135752.1 | NP_001129224.1 | 7 | 1138 | ||
| NM_007265.2 | NP_009196.1 | 7 | 1138 | |||
| NR_024203.1 | 5 | 727 | ||||
| GenBank mRNA | AK022616.1 | - | 7 | 1086 | ||
| AK315711.1 | - | 7 | 1111 | |||
| BC000721.2 | - | 7 | 1102 | 3504956 | ||
| CR457407.1 | - | 6 | 895 | |||
| D88208.1 | - | 7 | 1102 | |||

ECD
Genomic Map