siRNA Details
| Gene Symbol: MGAT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160154.1 | NP_001153626.1 | 5, 6 | 496 | ||
| NM_012214.2 | NP_036346.1 | 8, 9 | 1080 | |||
| GenBank mRNA | AB000616.1 | - | 7, 8 | 901 | ||
| AK291533.1 | - | 7, 8 | 890 | |||
| AK304393.1 | - | 5, 6 | 496 | |||
| AK314024.1 | - | 8, 9 | 1040 | |||
| BC127107.1 | - | 7, 8 | 978 | 40124731 | ||
| BC127108.1 | - | 7, 8 | 978 | 40124741 | ||

MGAT4A
Genomic Map