siRNA Details
| Gene Symbol: TMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127198.2 | NP_001120670.1 | 9 | 1310 | ||
| NM_001321185.1 | NP_001308114.1 | 9 | 1263 | |||
| NM_007267.7 | NP_009198.4 | 9 | 1214 | |||
| XM_005256995.1 | XP_005257052.1 | 9 | 1339 | |||
| XM_011524255.1 | XP_011522557.1 | 8 | 1241 | |||
| XM_011524256.1 | XP_011522558.1 | 8 | 1115 | |||
| XM_011524257.2 | XP_011522559.1 | 3 | 359 | |||
| XM_011524258.1 | XP_011522560.1 | 8 | 1241 | |||
| XM_017024107.1 | XP_016879596.1 | 9 | 1288 | |||
| XM_017024108.1 | XP_016879597.1 | 9 | 1241 | |||
| XM_017024109.1 | XP_016879598.1 | 7 | 1030 | |||
| GenBank mRNA | AB002405.2 | - | 1 | 22 | ||
| AK021738.1 | - | 7 | 1071 | |||
| AK074065.1 | - | 9 | 1213 | |||
| AK096076.1 | - | 9 | 1246 | |||
| AK310734.1 | - | 8 | 1634 | |||
| AY057379.1 | - | 8 | 1053 | |||
| AY099356.1 | - | 10 | 1212 | |||
| AY099357.1 | - | 10 | 1212 | |||
| AY236496.1 | - | 8 | 1179 | |||
| BC007766.1 | - | 8 | 953 | |||
| BC018346.1 | - | 2 | 178 | |||
| BC023597.2 | - | 9 | 1194 | 4566166 | ||
| BC035648.1 | - | 2 | 234 | 5575949 | ||

TMC6
Genomic Map