siRNA Details
| Gene Symbol: CTRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007272.2 | NP_009203.2 | 3 | 212 | ||
| XM_011540550.1 | XP_011538852.1 | 3 | 212 | |||
| GenBank mRNA | AK289447.1 | - | 3 | 211 | ||
| BC015118.1 | - | 3 | 218 | 3950170 | ||
| BC059779.1 | - | 3 | 191 | |||
| S82198.1 | - | 3 | 189 | |||
| Y13697.1 | - | 2 | 141 | |||

CTRC
Genomic Map