siRNA Details
| Gene Symbol: COG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145036.1 | NP_001138508.1 | 16 | 2067 | ||
| NM_007357.2 | NP_031383.1 | 16 | 2070 | |||
| GenBank mRNA | AK295251.1 | - | 16 | 2077 | ||
| AK302288.1 | - | 8 | 1043 | |||
| AK302791.1 | - | 17 | 2254 | |||
| AK312678.1 | - | 16 | 1986 | |||
| BC014960.1 | - | 16 | 2033 | 4876271 | ||
| BC051906.1 | - | 16 | 1956 | 4541036 | ||
| Z34975.1 | - | 16 | 2014 | |||

COG2
Genomic Map