siRNA Details
| Gene Symbol: COG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145036.1 | NP_001138508.1 | 2 | 298 | ||
| NM_007357.2 | NP_031383.1 | 2 | 298 | |||
| GenBank mRNA | AK295251.1 | - | 2 | 305 | ||
| AK302791.1 | - | 2 | 249 | |||
| AK312678.1 | - | 2 | 214 | |||
| AL832190.1 | - | 2 | 265 | |||
| BC014960.1 | - | 2 | 261 | 4876271 | ||
| BC051906.1 | - | 2 | 187 | 4541036 | ||
| DB449302.1 | - | 2 | 296 | |||
| DC349417.1 | - | 2 | 297 | |||
| Z34975.1 | - | 2 | 242 | |||

COG2
Genomic Map