siRNA Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012128.3 | NP_036260.2 | 7 | 1095 | ||
| NR_024602.1 | 6 | 986 | ||||
| XM_011541015.2 | XP_011539317.1 | 7 | 914 | |||
| GenBank mRNA | AF127035.1 | - | 7 | 1079 | ||
| AK000072.1 | - | 7 | 1093 | |||
| AK293766.1 | - | 6 | 750 | |||
| AK307785.1 | - | 6 | 931 | |||
| AK307898.1 | - | 5 | 700 | |||
| AK314430.1 | - | 7 | 1091 | |||
| AY358470.1 | - | 7 | 1075 | |||
| BC113687.1 | - | 7 | 1090 | 8322739 | ||
| BC113689.1 | - | 7 | 1089 | 8322741 | ||

CLCA4
Genomic Map