siRNA Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145365.1 | NP_001138837.1 | 2 | 597 | ||
| NM_014897.2 | NP_055712.1 | 2 | 452 | |||
| NR_135579.1 | 3 | 721 | ||||
| XM_005257166.2 | XP_005257223.1 | 1 | 344 | |||
| GenBank mRNA | AB023141.1 | - | 2 | 452 | ||
| AK292667.1 | - | 2 | 587 | |||
| BC139779.1 | - | 1 | 307 | 40122232 | ||
| BC152440.1 | - | 2 | 452 | 8862563 | ||
| CR933680.1 | - | 2 | 448 | |||
| DN998934.1 | - | 2 | 604 | |||

ZNF652
Genomic Map