siRNA Details
| Gene Symbol: COBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278458.1 | NP_001265387.1 | 9 | 1147 | ||
| NM_001278460.1 | NP_001265389.1 | 7 | 1087 | |||
| NM_001278461.1 | NP_001265390.1 | 6 | 1047 | |||
| NM_014900.4 | NP_055715.3 | 6 | 1047 | |||
| GenBank mRNA | AB023194.1 | - | 6 | 1041 | ||
| AK002054.1 | - | 8 | 1028 | |||
| AK225849.1 | - | 7 | 944 | |||
| AK294937.1 | - | 9 | 1147 | |||
| BC071588.1 | - | 6 | 1049 | 30345650 | ||
| BX537877.1 | - | 2 | 2649 | |||
| BX649112.1 | - | 1 | 30 | |||

COBLL1
Genomic Map